Learn More
Lonza Walkersville hMSC Adipogenic Induction Kit
Catalog No. 5000496449
Encompass
hMSC Adipogenic Induction SingleQuots Kit is a component of the hMSC Adipogenic Differentiation BulletKitTM and is available separately Stem cells possess the unique ability to differentiate into multiple lineages of cells including adipocytes Lonza s hMSC Adipogenic Differentiation BulletKitTM Medium is a serum containing media system designed to induce adipogenic differentiation of human bone marrow derived mesenchymal stem cells into mature functionally active adipocytes in as few as 19 days Differentiation can then be validated by a variety of methods including the use of Lonza s AdipoRedTM Adipogenic Assay Reagent which provides a quantitative value related to the amount of adipogenic differentiation
Catalog No. 50-004-96449 Supplier Lonza Walkersville Supplier No. PT4135
hMSC Adipogenic Induction SingleQuots Kit is a component of the hMSC Adipogenic Differentiation BulletKitTM and is available separately Stem cells possess the unique ability to differentiate into multiple lineages of cells including adipocytes Lonza s hMSC Adipogenic Differentiation BulletKitTM Medium is a serum containing media system designed to induce adipogenic differentiation of human bone marrow derived mesenchymal stem cells into mature functionally active adipocytes in as few as 19 days Differentiation can then be validated by a variety of methods including the use of Lonza s AdipoRedTM Adipogenic Assay Reagent which provides a quantitative value related to the amount of adipogenic differentiation