Learn More
Promocell GmbH PromoCell hMSC-AT-c 500,000 Human Mesenchymal Stem Cells, derived from Adipose Tissue, cryopreserved
Catalog No. 50306085
Encompass
Mesenchymal Stem Cells (MSC), also termed Mesenchymal Stromal Cells, are multipotent cells that can differentiate into a variety of cell types and have the capacity for self renewal. MSC have been shown to differentiate in vitro or in vivo into adipocytes, chondrocytes, osteoblasts, myocytes, neurons, hepatocytes, and pancreatic islet cells. Optimized PromoCell media are available to support both the growth of MSC and their differentiation into several different lineages.PromoCell hMSC are harvested from normal human adipose tissue, bone marrow, and umbilical cord matrix (Wharton’s jelly) of individual donors.The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, and osteoblasts. Our hMSC show a verified marker expression profile that complies with ISCT* recommendations, providing well characterized cells.*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317
Catalog No. 50-306-085 Supplier Promocell GmbH Supplier No. C12977
Mesenchymal Stem Cells (MSC), also termed Mesenchymal Stromal Cells, are multipotent cells that can differentiate into a variety of cell types and have the capacity for self renewal. MSC have been shown to differentiate in vitro or in vivo into adipocytes, chondrocytes, osteoblasts, myocytes, neurons, hepatocytes, and pancreatic islet cells. Optimized PromoCell media are available to support both the growth of MSC and their differentiation into several different lineages.PromoCell hMSC are harvested from normal human adipose tissue, bone marrow, and umbilical cord matrix (Wharton’s jelly) of individual donors.The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, and osteoblasts. Our hMSC show a verified marker expression profile that complies with ISCT* recommendations, providing well characterized cells.*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317