Learn More
Lonza Walkersville hMSC Chondro Suppl Growth F
Catalog No. 5000496389
Encompass
Lonza s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation with the addition of TGF- beta 3 of human bone marrow derived mesenchymal stem cells into mature functionally active chondrocytes in as few as 14 days Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin
Catalog No. 50-004-96389 Supplier Lonza Walkersville Supplier No. PT4121
Lonza s hMSC Chondrogenic Differentiation BulletKit Medium is a serum containing medium designed to induce chondrogenic differentiation with the addition of TGF- beta 3 of human bone marrow derived mesenchymal stem cells into mature functionally active chondrocytes in as few as 14 days Differentiation can then be validated by a variety of methods including staining with alcian blue or safranin