Learn More
Lonza Walkersville Human Mesenchymal Stem Cell (hMSC) Osteogenic Differentiation Medium BulletKit
Catalog No. NC9915521
Encompass
Stem cells possess the unique ability to differentiate into multiple lineages of cells including osteoblasts. Lonza’s hMSC Osteogenic Differentiation BulletKit Medium is a serum containing medium designed to induce osteogenic differentiation of human bone marrow derived mesenchymal stem cells (hMSC) into mature, functionally active osteoblasts in as few as 14 days. Differentiation can then be validated by a variety of methods including the use of Lonza’s OsteoImage Bone Mineralization Reagent which provides a quantitative value related to the amount of osteogenic differentiation.Content:1 x hMSC Osteogenic Differentiation Basal Medium (PT-3924), 170 mL1 x hMSC Osteogenic Differentiation SingleQuots Supplements Kit (PT-4120)
Catalog No. NC9915521 Supplier Lonza Walkersville Supplier No. PT3002
Stem cells possess the unique ability to differentiate into multiple lineages of cells including osteoblasts. Lonza’s hMSC Osteogenic Differentiation BulletKit Medium is a serum containing medium designed to induce osteogenic differentiation of human bone marrow derived mesenchymal stem cells (hMSC) into mature, functionally active osteoblasts in as few as 14 days. Differentiation can then be validated by a variety of methods including the use of Lonza’s OsteoImage Bone Mineralization Reagent which provides a quantitative value related to the amount of osteogenic differentiation.Content:1 x hMSC Osteogenic Differentiation Basal Medium (PT-3924), 170 mL1 x hMSC Osteogenic Differentiation SingleQuots Supplements Kit (PT-4120)