Learn More
Lonza Walkersville Human Mesenchymal Stem Cell (hMSC) Adipogenic Differentiation Medium BulletKit
Catalog No. NC9985002
Encompass
Stem cells possess the unique ability to differentiate into multiple lineages of cells including adipocytes. Lonza’s hMSC Adipogenic Differentiation BulletKit Medium is a serum containing media system designed to induce adipogenic differentiation of bone marrow derived hMSC into mature, functionally active adipocytes in as few as 19 days. Differentiation can then be validated by a variety of methods including the use of Lonza’s AdipoRed Adipogenic Assay Reagent which provides a quantitative value related to the amount of adipogenic differentiation.Content:1 x hMSC Adipogenic Maintenance Medium (PT-3102A)1 x hMSC Adipogenic Induction Medium (PT-3102B)1 x hMSC Adipogenic Maintenance SingelQuot Kit (PT-4122)1 x hMSC Adipogenic Induction SingleQuot Kit (PT-4135)
Catalog No. NC9985002 Supplier Lonza Walkersville Supplier No. PT3004
Stem cells possess the unique ability to differentiate into multiple lineages of cells including adipocytes. Lonza’s hMSC Adipogenic Differentiation BulletKit Medium is a serum containing media system designed to induce adipogenic differentiation of bone marrow derived hMSC into mature, functionally active adipocytes in as few as 19 days. Differentiation can then be validated by a variety of methods including the use of Lonza’s AdipoRed Adipogenic Assay Reagent which provides a quantitative value related to the amount of adipogenic differentiation.Content:1 x hMSC Adipogenic Maintenance Medium (PT-3102A)1 x hMSC Adipogenic Induction Medium (PT-3102B)1 x hMSC Adipogenic Maintenance SingelQuot Kit (PT-4122)1 x hMSC Adipogenic Induction SingleQuot Kit (PT-4135)