Learn More
Lonza Walkersville hMSC - Human Mesenchymal Stem Cells
Catalog No. NC9685231
Encompass
Poietics Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are isolated from normal (non-diabetic) adult human bone marrow withdrawn from bilateral punctures of the posterior iliac crests of normal volunteers. Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been reported to differentiate down many different lineages including chondrogenic, osteogenic, adipogenic and neural. Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been cryopreserved after the second passage and are suggested to be used by no later than the fifth passage. Cryopreserved Poietics Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are guaranteed to have at least 0.75 million viable cells per vial, differentiate down the adipogenic, chondrogenic, and osteogenic liniages when cultured in the recommended differentiation medium, to express CD29, CD44, CD105, and CD166 and to not express CD14, CD34, and CD45.
Catalog No. NC9685231 Supplier Lonza Walkersville Supplier No. PT2501
Poietics Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are isolated from normal (non-diabetic) adult human bone marrow withdrawn from bilateral punctures of the posterior iliac crests of normal volunteers. Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been reported to differentiate down many different lineages including chondrogenic, osteogenic, adipogenic and neural. Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) have been cryopreserved after the second passage and are suggested to be used by no later than the fifth passage. Cryopreserved Poietics Normal Human Bone Marrow Derived Mesenchymal Stem Cells (hMSC) are guaranteed to have at least 0.75 million viable cells per vial, differentiate down the adipogenic, chondrogenic, and osteogenic liniages when cultured in the recommended differentiation medium, to express CD29, CD44, CD105, and CD166 and to not express CD14, CD34, and CD45.