Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza Walkersville Human Cardiac Microvascular Endothelial Cells, cryopreserved ampule, ≥ 500,000 cells, Passage 3, EGM™-2 MV Growth Medium (CC-3202)

Catalog No. NC3446055
Encompass

Human Cardiac Microvascular Endothelial Cells (HMVEC-C) are primary cells derived from human heart tissue and are specifically tailored for cardiovascular research. These cryopreserved cells are isolated from both normal and diabetic donors, ensuring a robust platform for various experimental applications. HMVEC-C are subjected to extensive quality testing, guaranteeing their performance across multiple population doublings, while being free from common contaminants.

  • Cell type: Primary human cardiac microvascular endothelial cells
  • Tissue origin: Heart
  • Species: Human
  • Donor status: Normal and diabetic donors
  • Format: Cryopreserved ampule
  • Quantity: ≥ 500,000 cells per ampule
  • Passage number at shipment: 3
  • Guaranteed population doublings: 10 (primary), 15 (secondary)
  • Positive for acetylated low density lipoprotein uptake
  • Quality assurance: Certificate of Analysis provided

Catalog No. NC3446055 Supplier Lonza Walkersville Supplier No. CC7030
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Human Cardiac Microvascular Endothelial Cells (HMVEC-C) are primary cells derived from human heart tissue and are specifically tailored for cardiovascular research. These cryopreserved cells are isolated from both normal and diabetic donors, ensuring a robust platform for various experimental applications. HMVEC-C are subjected to extensive quality testing, guaranteeing their performance across multiple population doublings, while being free from common contaminants.

  • Cell type: Primary human cardiac microvascular endothelial cells
  • Tissue origin: Heart
  • Species: Human
  • Donor status: Normal and diabetic donors
  • Format: Cryopreserved ampule
  • Quantity: ≥ 500,000 cells per ampule
  • Passage number at shipment: 3
  • Guaranteed population doublings: 10 (primary), 15 (secondary)
  • Positive for acetylated low density lipoprotein uptake
  • Quality assurance: Certificate of Analysis provided

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.