Learn More
Lonza Walkersville HMVEC-L – Human Lung Microvascular Endothelial Cells
Catalog No. NC1949892
Encompass
Lonza's Lung Microvascular Endothelial Cells include cells isolated from small vessels within normal lung tissue that are cultured to be ≥90% pure and specifically enriched for lung LECs and BECs. Lung microvascular cells, CC-2527 (HMVEC-L), are a mixed population of both blood (HMVEC-LBl) and lymphatic (HMVEC-LLy) endothelial cells.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate of Analysis is provided for each cell lot purchased Applications:OncologyTumorigenesisDrug DevelopmentCirculatory PhysiologyWound HealingLymphangiogenesisImmunologyBasic ResearchContent:1 x Cryopreserved ampule of HMVEC-L containing ≥ 500,000 cells
Catalog No. NC1949892 Supplier Lonza Walkersville Supplier No. CC2527
Lonza's Lung Microvascular Endothelial Cells include cells isolated from small vessels within normal lung tissue that are cultured to be ≥90% pure and specifically enriched for lung LECs and BECs. Lung microvascular cells, CC-2527 (HMVEC-L), are a mixed population of both blood (HMVEC-LBl) and lymphatic (HMVEC-LLy) endothelial cells.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate of Analysis is provided for each cell lot purchased Applications:OncologyTumorigenesisDrug DevelopmentCirculatory PhysiologyWound HealingLymphangiogenesisImmunologyBasic ResearchContent:1 x Cryopreserved ampule of HMVEC-L containing ≥ 500,000 cells