Learn More
New England Biolabs, Inc. Monarch® HMW gDNA Tissue Lysis Buffer - 62 ml
SDP
Catalog No. 502068312
Encompass
The Monarch HMW gDNA Tissue Lysis Buffer, supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue, is a highly effective lysis buffer for tissue and bacteria samples. Its composition ensures immediate and complete inactivation of nucleases in the samples at the start of the lysis.
  • Supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue
  • This lysis buffer enables rapid and effective lysis of various tissue and bacteria samples
  • Offers maximal protection against nucleases allowing for the isolation of intact HMW DNA
  • Supplied as one bottle of 62 ml
Catalog No. 50-206-8312 Supplier New England Biolabs, Inc. Supplier No. T3061L
The Monarch HMW gDNA Tissue Lysis Buffer, supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue, is a highly effective lysis buffer for tissue and bacteria samples. Its composition ensures immediate and complete inactivation of nucleases in the samples at the start of the lysis.
  • Supplied as a component of the Monarch HMW DNA Extraction Kit for Tissue
  • This lysis buffer enables rapid and effective lysis of various tissue and bacteria samples
  • Offers maximal protection against nucleases allowing for the isolation of intact HMW DNA
  • Supplied as one bottle of 62 ml