Learn More
Aviva Systems Biology Corp Hmx3 Antibody - middle region (ARP47785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hmx3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Nkx-5.1, Nkx5-1, Nkx5.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-285-328