Learn More
Medchemexpress LLC HN37 | 1821222-10-9 | 99.5% | 340.39 g/mol | C20H21FN2O2 | 5 MG
SDP
Catalog No. 5000221416
Encompass_Preferred
HN37 is a small-molecule research compound that functions as a potent activator of the KCNQ2 potassium channel. It contains an alkyne moiety suitable for click-chemistry derivatization and is intended for in vitro pharmacology and chemical biology applications.
  • Potent KCNQ2 activation with EC50 of 37 nM.
  • Alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition.
  • High reported purity for reproducible assay performance.
  • Soluble in DMSO at 50 mg/mL for preparation of concentrated stocks.
  • Available in multiple milligram quantities for research use.
Catalog No. 50-002-21416 Supplier Medchemexpress LLC Supplier No. HY1450165MG
HN37 is a small-molecule research compound that functions as a potent activator of the KCNQ2 potassium channel. It contains an alkyne moiety suitable for click-chemistry derivatization and is intended for in vitro pharmacology and chemical biology applications.
  • Potent KCNQ2 activation with EC50 of 37 nM.
  • Alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition.
  • High reported purity for reproducible assay performance.
  • Soluble in DMSO at 50 mg/mL for preparation of concentrated stocks.
  • Available in multiple milligram quantities for research use.