Learn More
Medchemexpress LLC HN37 | 1821222-10-9 | 99.5% | C20H21FN2O2 | 50 MG
SDP
Catalog No. 5000355355
Encompass_Preferred
HN37 is a potent and chemically stable antiepileptic agent candidate with an EC50 of 37 nM for KCNQ2. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. In vitro, HN37 is a potent neuronal Kv7 activator with subtype selectivity similar to RTG.
  • Potent and chemically stable antiepileptic agent candidate
  • EC50 of 37 nM for KCNQ2
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Potent neuronal Kv7 activator with subtype selectivity similar to RTG
Catalog No. 50-003-55355 Supplier Medchemexpress LLC Supplier No. HY14501650MG
HN37 is a potent and chemically stable antiepileptic agent candidate with an EC50 of 37 nM for KCNQ2. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. In vitro, HN37 is a potent neuronal Kv7 activator with subtype selectivity similar to RTG.
  • Potent and chemically stable antiepileptic agent candidate
  • EC50 of 37 nM for KCNQ2
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Potent neuronal Kv7 activator with subtype selectivity similar to RTG