Learn More
Eurofins Discoverx PrecisION hNav1.1 Recombinant Stable Cell Line, 2 vials. (CYL3009)
Catalog No. 502004796
Encompass
The PrecisION hNav1.1 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Nav1.1 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Nav1.1 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.
Catalog No. 50-200-4796 Supplier Eurofins Discoverx Supplier No. CYL3009
Add to Cart
Add to Cart
Request a Quote
The PrecisION hNav1.1 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Nav1.1 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Nav1.1 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.