Learn More
Med Vet International Miltex Knife Handle, 5in, #3, Holds Blade #'S 10 Through 15C, 4-7R
Catalog No. 502829517
Encompass_Preferred
Miltex Knife Handle, 5in, #3, Holds Blade #'s 10 Through 15C, 4-7R
Catalog No. 50-282-9517 Supplier Med Vet International Supplier No. MIL47R
Miltex Knife Handle, 5in, #3, Holds Blade #'s 10 Through 15C, 4-7R