Learn More
Med Vet International Miltex Knife Handle, #3, Fits #10-15 Blades,Eg4-7
Catalog No. 502829742
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These is a Miltex Knife Handle. Scalpels or Scalpel Handles are intended to be use with surgical blades for tissue separation and other procedure that require a sharp surgical blade to puncture or cut. Specifications: #3, Fits #10-15 Blades
Catalog No. 50-282-9742 Supplier Med Vet International Supplier No. MILEG47
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These is a Miltex Knife Handle. Scalpels or Scalpel Handles are intended to be use with surgical blades for tissue separation and other procedure that require a sharp surgical blade to puncture or cut. Specifications: #3, Fits #10-15 Blades