Learn More
Cell Biolabs Inc OxiSelect™ HNE Adduct Competitive ELISA Kit, Trial Size, 32 assays
Catalog No. 50673746
Encompass_Preferred
4-Hydroxynonenal (HNE) is a common and relatively stable byproduct of lipid peroxidation. It is generated from the breakdown of lipid peroxides which are difficult to measure directly. Therefore HNE is a well-published marker of lipid peroxidation. Cell Biolabs' OxiSelect™ HNE Adduct Competitive ELISA Kit provides rapid detection of malondialdehyde-protein adducts in cell and tissue lysates, plasma, serum, or purified proteins. Each trial size kit provides sufficient reagents for 32 assays including standards and unknown samples.
Catalog No. 50-673-746 Supplier Cell Biolabs Inc Supplier No. STA838T
Add to Cart
Add to Cart
4-Hydroxynonenal (HNE) is a common and relatively stable byproduct of lipid peroxidation. It is generated from the breakdown of lipid peroxides which are difficult to measure directly. Therefore HNE is a well-published marker of lipid peroxidation. Cell Biolabs' OxiSelect™ HNE Adduct Competitive ELISA Kit provides rapid detection of malondialdehyde-protein adducts in cell and tissue lysates, plasma, serum, or purified proteins. Each trial size kit provides sufficient reagents for 32 assays including standards and unknown samples.