Learn More
Teknova HNE Buffer, 1000mL, Sterile.
SDP
Catalog No. 502745962
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-274-5962 Supplier Teknova Supplier No. 21509
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.