Learn More
Promocell GmbH PromoCell HNEpC-c 500,000 Human Nasal Epithelial Cells, cryopreserved
Catalog No. 50306044
Encompass
Primary Human Nasal Epithelial Cells (HNEpC) are isolated from normal human nasal mucosa and stain positive for cytokeratin.The epithelium in the nasal cavity cleans, humidifies, and warms inhaled air. In addition, it produces mucus, which binds particles that are subsequently transported to the pharynx by cilia on the epithelial cells. HNEpC are useful for in vitro studies of these processes.Our HNEpC are now also available from HLA-typed donors.
Catalog No. 50-306-044 Supplier Promocell GmbH Supplier No. C12620
Primary Human Nasal Epithelial Cells (HNEpC) are isolated from normal human nasal mucosa and stain positive for cytokeratin.The epithelium in the nasal cavity cleans, humidifies, and warms inhaled air. In addition, it produces mucus, which binds particles that are subsequently transported to the pharynx by cilia on the epithelial cells. HNEpC are useful for in vitro studies of these processes.Our HNEpC are now also available from HLA-typed donors.