Learn More
Aviva Systems Biology Corp HNF1B Antibody - N-terminal region (ARP32999_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32999_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNF1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNF1B. Alternate names: FJHN, HNF1beta, HNF2, LFB3, MODY5, TCF2, VHNF1, HPC11, LF-B3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54056