Learn More
Abnova™ HNF1B/CEN17q FISH Probe
Catalog No. 89999057 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 uL
Catalog No. Quantity
89-999-057 100 uL
1 options
Catalog No. 89-999-057 Supplier Abnova™ Supplier No. FG0230
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
Quantity 100 uL
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe