Learn More
Aviva Systems Biology Corp HNF4A Antibody - middle region (ARP45595_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45595_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNF4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HNF4A. Alternate names: FLJ39654, HNF4, HNF4a7, HNF4a8, HNF4a9, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14, HNF4alphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59058