Learn More
Aviva Systems Biology Corp HNF4A Antibody - N-terminal region (ARP31946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNF4A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human HNF4A. Alternate names: HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54387