Learn More
Aviva Systems Biology Corp HNF4G Antibody - N-terminal region (ARP45615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNF4G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNF4G. Alternate names: NR2A2, NR2A3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-53449