Learn More
Abcam HNHA, HDAC inhibitor, 25MG
Catalog No. 502706750
Encompass_Preferred
MW 303.4 Da, Purity >98%. Potent, cell-permeable HDAC inhibitor (IC₅₀ = 100 nM). Induces histone hyperacetylation and p21 transcription with concomitant inhibition of cell cycle progression. Arrests cell cycle at G₁/S phase in cancer cells. Antitumor and antiangiogenic agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6750 Supplier Abcam Supplier No. AB14126225MG
MW 303.4 Da, Purity >98%. Potent, cell-permeable HDAC inhibitor (IC₅₀ = 100 nM). Induces histone hyperacetylation and p21 transcription with concomitant inhibition of cell cycle progression. Arrests cell cycle at G₁/S phase in cancer cells. Antitumor and antiangiogenic agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement