Learn More
Enzo Life Sciences Hydroxy-2-naphthalenylmethylphosphonic acid (25mg). CAS: 132541-52-7
SDP
Supplier:  Enzo Life Sciences BMLEI2470025
Encompass_Preferred
HNMPA is a membrane impermeable inhibitor of insulin receptor tyrosine kinase (IC50 = 100µM); in rat adipocytes (IC50 = 10µM). Inhibits serine and tyrosine autophosphorylation by the human insulin receptor. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml), DMSO (25ml/ml) or 5% Sodium bicarbonate (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0634