Learn More
Aviva Systems Biology Corp HNMT Antibody - N-terminal region (ARP59621_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59621_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNMT. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HNMT. Alternate names: HMT, HNMT-S1, HNMT-S2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-759