Learn More
Medchemexpress LLC Hnpmi | 1980848-48-3 | 99.6% | 360.41 | 50 MG
SDP
Catalog No. 5000392820
Encompass_Preferred
HNPMI is an EGFR inhibitor that exhibits cytotoxic effects on tumor cells. It downregulates protein levels of osteopontin, survivin, and cathepsin S, inducing apoptosis. Additionally, HNPMI modulates BCL-2/BAX and p53 in CRC cell lines to hinder tumorigenesis.
  • Inhibitor of EGFR.
  • Cytotoxic effects on tumor cells.
  • Downregulates osteopontin, survivin, and cathepsin S.
  • Induces apoptosis.
  • Regulates BCL-2/BAX and p53 in CRC cell lines.
Catalog No. 50-003-92820 Supplier Medchemexpress LLC Supplier No. HY16214850MG
HNPMI is an EGFR inhibitor that exhibits cytotoxic effects on tumor cells. It downregulates protein levels of osteopontin, survivin, and cathepsin S, inducing apoptosis. Additionally, HNPMI modulates BCL-2/BAX and p53 in CRC cell lines to hinder tumorigenesis.
  • Inhibitor of EGFR.
  • Cytotoxic effects on tumor cells.
  • Downregulates osteopontin, survivin, and cathepsin S.
  • Induces apoptosis.
  • Regulates BCL-2/BAX and p53 in CRC cell lines.