Learn More
Aviva Systems Biology Corp HNRNPA2B1 Antibody - N-terminal region (ARP41035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRNPA2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNRNPA2B1. Alternate names: DKFZp779B0244, FLJ22720, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-53266