Learn More
Aviva Systems Biology Corp HNRNPC Antibody - middle region (ARP40517_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40517_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRNPC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HNRNPC. Alternate names: C1, C2, HNRNP, HNRPC, MGC104306, MGC105117, MGC117353, MGC131677, SNRPCApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57498