Learn More
Aviva Systems Biology Corp Hnrnpc Antibody - C-terminal region (ARP40516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hnrnpc. It was validated on Western Blot. . Alternate names: AL022939, D14Wsu171e, Hnrpc, Hnrpc1, Hnrpc2, hnRNPC1, hnRNPC2, hnrnp-CApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-284-877