Learn More
Aviva Systems Biology Corp HNRNPF Antibody - C-terminal region (ARP40566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRNPF. It was validated on Western Blot. . Alternate names: HNRPF, MGC110997, OK/SW-cl.23, mcs94-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-880