Learn More
Aviva Systems Biology Corp HNRNPR Antibody - N-terminal region (ARP40605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRNPR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNRNPR. Alternate names: FLJ25714, HNRPR, hnRNP-R, hnRNP RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57544