Learn More
Aviva Systems Biology Corp HNRNPU Antibody - N-terminal region (ARP40519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRNPU. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNRNPU. Alternate names: HNRPU, SAF-A, U21.1, hnRNP UApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57499