Learn More
Aviva Systems Biology Corp HNRPA1 Antibody - C-terminal region (ARP40384_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40384_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPA1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human HNRPA1. Alternate names: HNRPA1, HNRPA1L3, hnRNP A1, hnRNP-A1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57423