Learn More
Aviva Systems Biology Corp HNRPAB Antibody - C-terminal region (ARP40514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPAB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HNRPAB. Alternate names: ABBP1, FLJ40338, HNRPABApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-876