Learn More
Aviva Systems Biology Corp HNRPC Antibody - N-terminal region (ARP41037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNRPC. Alternate names: C1, C2, HNRNP, MGC104306, MGC105117, MGC117353, MGC131677, SNRPC, hnRNPC, HNRPCApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57699