Learn More
Aviva Systems Biology Corp HNRPD Antibody - N-terminal region (ARP40239_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40239_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPD. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HNRPD. Alternate names: P37, AUF1, AUF1A, HNRPD, hnRNPD0ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57361