Learn More
Aviva Systems Biology Corp HNRNPDL Antibody - C-terminal region (ARP40586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPDL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HNRPDL. Alternate names: HNRNP, JKTBP, JKTBP2, laAUF1, HNRPDLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53211