Learn More
Aviva Systems Biology Corp HNRPL Antibody - N-terminal region (ARP40368_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40368_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HNRPL. Alternate names: HNRPL, hnRNP-L, P/OKcl.14ApplicationIF, IP, WB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57413