Learn More
Aviva Systems Biology Corp HNRPLL Antibody - N-terminal region (ARP41102_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41102_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPLL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HNRPLL. Alternate names: SRRF, hnRNPLL, HNRPLLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-014-53272