Learn More
Aviva Systems Biology Corp HNRPLL Antibody - N-terminal region (ARP41103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPLL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HNRPLL. Alternate names: SRRF, hnRNPLL, HNRPLLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53273