Learn More
Aviva Systems Biology Corp HNRPUL1 Antibody - C-terminal region (ARP40702_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40702_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HNRPUL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human HNRPUL1. Alternate names: E1BAP5, E1B-AP5, HNRPUL1ApplicationIF, IP, WB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57595