Learn More
Enzo Life Sciences HO-1 monoclonal antibody (HO-1-2) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIOSA111F
Encompass_Preferred
Clone: HO-1-2. Host: Mouse. Isotype: IgG2b. Immunogen: Native rat HO-1. UniProt: P06762. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog. Applications: Flow Cytometry, IHC, WB. Recommended dilutions: Flow Cytometry (1:100). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-003