Learn More
Enzo Life Sciences HO-1 (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP732F
Encompass_Preferred
Alternative name: HMOX1, HSP32, Heat shock protein 32, Heme oxygenase 1. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~32kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 2% glycerol, 450mM sodium chloride, 1mM DTT, and 0.1mM PMSF. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Antigen microarray.
Catalog No. 50-200-7626