Learn More
Enzo Life Sciences HO-1 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS800
Encompass_Preferred
Highly sensitive HO-1 ELISA kit enabling detection of HO-1 in cell lysates and tissue extracts in < 2.5 hours. The HO-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 2.5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative Name: Heme oxygenase 1. Sensitivity: 0.78 ng/ml (range 0.78 - 25 ng/ml). Assay Time: <2.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2
Catalog No. 50-659-137