Learn More
Enzo Life Sciences HO-1 monoclonal antibody (HO-1-1) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6630200
Encompass_Preferred
Clone: HO-1-1. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: Flow Cytometry, ICC, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3189