Learn More
Enzo Life Sciences HO-1 monoclonal antibody (HO-1-2) (1mg)
SDP
Supplier:  Enzo Life Sciences ADIOSA111J
Encompass_Preferred
Clone: HO-1-2. Host: Mouse. Isotype: IgG2b. Immunogen: Native rat HO-1. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog. Applications: Flow Cytometry, IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7362