Learn More
Enzo Life Sciences HO-1 monoclonal antibody (HO-1-2) (FITC conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS687FI0200
Encompass_Preferred
Clone: HO-1-2. Isotype: IgG2b. Immunogen: Native rat HO-1. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-3220