Learn More
Enzo Life Sciences HO 1 monoclonal antibody (HO-1-1) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIOSA110D
Encompass_Preferred
Clone: HO-1-1. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1. UniProt: P09601. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: Flow Cytometry, ICC, IHC (PS), WB. Recommended dilutions: Flow Cytometry (10µg/ml). Western Blot (1:1,000 ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-666-996