Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ho-1 protein, mouse (P. pastoris, His) | ≥90.0% | 20UG
SDP

Catalog No. 5000176616
Encompass_Preferred

Recombinant His-tagged mouse heme oxygenase-1 (HO-1) produced in Pichia pastoris and supplied lyophilized with stabilizing excipients for biochemical and enzymatic studies. The protein shows glycosylated forms on SDS-PAGE, has low endotoxin, and is intended for use in heme metabolism and oxidative stress research.

  • Recombinant His-tagged protein expressed in P. pastoris.
  • Observed molecular weight ~31-34 kDa due to glycosylation.
  • Endotoxin level <1 EU/μg, suitable for sensitive assays.
  • Lyophilized formulation with 6% trehalose for stability.
  • Store at -20°C; reconstituted stability 4°C for 1 week.
  • Suitable for enzymatic assays, biochemical characterization, and signaling studies.
  • Supplied as a 20 μg vial for small-scale experiments.

Catalog No. 50-001-76616 Supplier Medchemexpress LLC Supplier No. HYP70049520UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant His-tagged mouse heme oxygenase-1 (HO-1) produced in Pichia pastoris and supplied lyophilized with stabilizing excipients for biochemical and enzymatic studies. The protein shows glycosylated forms on SDS-PAGE, has low endotoxin, and is intended for use in heme metabolism and oxidative stress research.

  • Recombinant His-tagged protein expressed in P. pastoris.
  • Observed molecular weight ~31-34 kDa due to glycosylation.
  • Endotoxin level <1 EU/μg, suitable for sensitive assays.
  • Lyophilized formulation with 6% trehalose for stability.
  • Store at -20°C; reconstituted stability 4°C for 1 week.
  • Suitable for enzymatic assays, biochemical characterization, and signaling studies.
  • Supplied as a 20 μg vial for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.